Fenoprofen calcium salt dihydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-(3-Phenoxyphenyl)propionic Acid Calcium Salt
F0372-25G
TCF0372-025GEA
216
USD
TCF0372-025G
Fenoprofen calcium salt dihydrate ≥98.0% (by titrimetric analysis)
Fenoprofen calcium salt dihydrate
CAS Number: 71720-56-4
MDL Number: MFCD00242722
Molecular Formula: C15H14O3
Molecular Weight: 522.61
Purity/Analysis Method:
Form: Crystal
Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-rheumatoid Arthritis Ingredients
Other Product Names / Synonyms: 2-(3-Phenoxyphenyl)propionic Acid Calcium Salt
Formula:
C₁₅H₁₄O₃ MW: 558,63 g/mol |
MDL Number:
MFCD00242722 CAS Number: 71720-56-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...