TCIAA1334

A1334-25ML
TCA1334-025MLEA 244.91 USD
TCA1334-025ML
TCIAA1334
CAS Number: 76497-39-7
MDL Number: MFCD00038563
Molecular Formula: C6H10O3S
Molecular Weight: 162.20
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Boiling point (°C): 132
Melting point (°C): 5
Flash Point (°C): 162
Specific Gravity (20/20): 1.18
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR