1-Fluoro-2,4,6-trimethylpyridiniumtetrafluoroborate ≥95.0% (by titrimetric analysis)

Synonyms: 1-Fluoro-2,4,6-trimethylpyridinium Tetrafluoroborate

F0346-5G
TCF0346-005GEA 184.48 USD
TCF0346-005G
1-Fluoro-2,4,6-trimethylpyridiniumtetrafluoroborate ≥95.0% (by titrimetric analysis)
1-Fluoro-2,4,6-trimethylpyridinium tetrafluoroborate

[Fluorinating Reagent]
CAS Number: 109705-14-8
MDL Number: MFCD00153182
Molecular Formula: C8H11BF5N
Molecular Weight: 226.99
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Melting point (°C): 186

Formula: C₈H₁₁BF₅N
MW: 226.98 g/mol
Melting Pt: 186 °C
MDL Number: MFCD00153182
CAS Number: 109705-14-8
https://us.vwr.com/stibo/web/std.lang.all/89/37/28968937.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR