1-Fluoro-2,6-dichloropyridiniumtetrafluoroborate ≥96.0% (by titrimetric analysis)

Synonyms: 2,6-Dichloro-N-fluoropyridinium tetrafluoroborate

F0344-5G F0344-25G
TCF0344-025GEA 487.29 USD
TCF0344-025G TCF0344-005G
1-Fluoro-2,6-dichloropyridiniumtetrafluoroborate ≥96.0% (by titrimetric analysis)
1-Fluoro-2,6-dichloropyridinium tetrafluoroborate

[Fluorinating Reagent]
CAS Number: 140623-89-8
MDL Number: MFCD00153157
Molecular Formula: C5H3BCl2F5N
Molecular Weight: 253.79
Purity/Analysis Method: 96.0% (T)
Form: Crystal
Melting point (°C): 245


Product Categories: Synthetic Organic Chemistry, Fluorinating Agents
Other Product Names / Synonyms: 2,6-Dichloro-1-fluoropyridinium Tetrafluoroborate

Formula: C₅H₃BCl₂F₅N
Melting Pt: 245 °C
MDL Number: MFCD00153157
CAS Number: 140623-89-8
https://us.vwr.com/stibo/web/std.lang.all/89/36/28968936.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR