L(+)-Fructose ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: L(+)-Levulose

F0317-1G F0317-100MG
TCF0317-1GEA 975.69 USD
TCF0317-1G TCF0317-100MG
L(+)-Fructose ≥95.0% (by HPLC)
L(+)-Fructose

CAS Number: 7776-48-9
MDL Number: MFCD05662378
Molecular Formula: C6H12O6
Molecular Weight: 180.16
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 89 deg (C=5, H2O)

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05662378
CAS Number: 7776-48-9
https://us.vwr.com/stibo/web/std.lang.all/89/10/28968910.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR