Cyclopentenylferrocene
Supplier: TCI America
Synonyms:
Cyclopentenylbis(cyclopentadienyl) Iron
, Ferrocenecyclopenten-1-yl
F0313-25G
TCF0313-025GEA
174.5
USD
TCF0313-025G
Cyclopentenylferrocene
Cyclopentenylferrocene
CAS Number: 12260-67-2
MDL Number: MFCD00191436
Molecular Formula: C15H16Fe
Molecular Weight: 252.14
Form: Crystal
Color: Yellow
Melting point (°C): 66
Other Product Names / Synonyms: Cyclopentenylbis(cyclopentadienyl) Iron, Ferrocenecyclopenten-1-yl
Formula:
C₁₅H₁₆Fe MW: 252.14 g/mol Melting Pt: 66 °C |
MDL Number:
MFCD00191436 CAS Number: 12260-67-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...