2-Methylcyclohexyl acetate ≥97.0%

Supplier: TCI America

Synonyms: Acetic Acid 2-Methylcyclohexyl Ester

A1329-25ML
TCA1329-025MLEA 114 USD
TCA1329-025ML
2-Methylcyclohexyl acetate ≥97.0%
2-Methylcyclohexyl acetate

CAS Number: 5726-19-2
MDL Number: MFCD00021390
Molecular Formula: C9H16O2
Molecular Weight: 156.23
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 182
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Acetic Acid 2-Methylcyclohexyl Ester

Formula: C₉H₁₆O₂
MW: 156.23 g/mol
Boiling Pt: 182 °C
Density: 0.95 (20 °C)
MDL Number: MFCD00021390
CAS Number: 5726-19-2
https://us.vwr.com/stibo/web/std.lang.all/92/44/28929244.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR