(Ferrocenylmethyl)dodecyldimethylammonium bromide ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Dodecyl(ferrocenylmethyl)dimethylammonium Bromide

F0286-5G
TCF0286-005GEA 302 USD
TCF0286-005G
(Ferrocenylmethyl)dodecyldimethylammonium bromide ≥97.0% (by titrimetric analysis)
(Ferrocenylmethyl)dodecyldimethylammonium bromide

CAS Number: 98778-40-6
MDL Number: MFCD00144308
Molecular Formula: C25H42BrFeN
Molecular Weight: 492.37
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 137


Other Product Names / Synonyms: Dodecyl(ferrocenylmethyl)dimethylammonium Bromide

Formula: C₂₅H₄₂BrFeN
MW: 492.37 g/mol
Melting Pt: 137 °C
MDL Number: MFCD00144308
CAS Number: 98778-40-6
https://us.vwr.com/stibo/web/std.lang.all/88/82/28968882.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR