6-Fluoro-2-methyl-4-chromanone ≥97.0%

Supplier: TCI America

F0271-5G
TCF0271-005GEA 32.43 USD
TCF0271-005G
6-Fluoro-2-methyl-4-chromanone ≥97.0%
6-Fluoro-2-methyl-4-chromanone

CAS Number: 88754-96-5
MDL Number: MFCD00059072
Molecular Formula: C10H9FO2
Molecular Weight: 180.18
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 71

Formula: C₁₀H₉FO₂
Melting Pt: 71 °C
MDL Number: MFCD00059072
CAS Number: 88754-96-5
https://us.vwr.com/stibo/web/std.lang.all/88/69/28968869.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR