4-Fluoro-4'-hydroxybenzophenone ≥98.0%

Supplier: TCI America

Synonyms: (4-Fluorophenyl)(4-hydroxyphenyl)methanone

F0269-10G
TCF0269-010GEA 57.51 USD
TCF0269-010G
4-Fluoro-4'-hydroxybenzophenone ≥98.0%
4-Fluoro-4'-hydroxybenzophenone

CAS Number: 25913-05-7
MDL Number: MFCD00011484
Molecular Formula: C13H9FO2
Molecular Weight: 216.21
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 168

Formula: C₁₃H₉FO₂
MW: 216.2078 g/mol
Melting Pt: 168 °C
MDL Number: MFCD00011484
CAS Number: 25913-05-7
https://us.vwr.com/stibo/web/std.lang.all/88/68/28968868.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR