2-Furfurylthio-3-methylpyrazine ≥95.0%

Supplier: TCI America

F0222-5G
TCF0222-005GEA 306 USD
TCF0222-005G
2-Furfurylthio-3-methylpyrazine ≥95.0%
2-Furfurylthio-3-methylpyrazine

CAS Number: 59303-07-0
MDL Number: MFCD00055031
Molecular Formula: C10H10N2OS
Molecular Weight: 206.26
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.22

Formula: C₁₀H₁₀N₂OS
Density: 1,22 (20 °C)
MDL Number: MFCD00055031
CAS Number: 59303-07-0

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR