Butyl fluoroacetate ≥97.0%

Supplier: TCI America

Synonyms: Fluoroacetic Acid Butyl Ester

F0215-25ML
TCF0215-025MLEA 161.34 USD
TCF0215-025ML
Butyl fluoroacetate ≥97.0%
Butyl fluoroacetate

CAS Number: 371-49-3
MDL Number: MFCD00042194
Molecular Formula: C6H11FO2
Molecular Weight: 134.15
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 154
Specific Gravity (20/20): 1.02


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Fluoroacetic Acid Butyl Ester

Formula: C₆H₁₁FO₂
MW: 134.15 g/mol
Boiling Pt: 154 °C
Density: 1.02 (20 °C)
MDL Number: MFCD00042194
CAS Number: 371-49-3
https://us.vwr.com/stibo/web/std.lang.all/88/21/28968821.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR