2-Methylpentyl formate ≥90.0%

Supplier: TCI America

Synonyms: Formic Acid 2-Methylpentyl Ester

F0174-5ML
TCF0174-005MLEA 74.06 USD
TCF0174-005ML
2-Methylpentyl formate ≥90.0%
2-Methylpentyl formate

CAS Number: 381670-34-4
MDL Number: MFCD00059817
Molecular Formula: C7H14O2
Molecular Weight: 130.19
Purity/Analysis Method: 90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: Formic Acid 2-Methylpentyl Ester

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 148 °C
Density: 0.88 (20 °C)
MDL Number: MFCD00059817
CAS Number: 381670-34-4

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR