2-Propoxyethylamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Aminoethyl Propyl Ether2-AminoethylpropyletherO-Propylethanolamine

A1325-25ML A1325-5ML
TCA1325-25MLEA 472.64 USD
TCA1325-25ML TCA1325-005ML
2-Propoxyethylamine ≥97.0% (by GC, titration analysis)
2-Propoxyethylamine

CAS Number: 42185-03-5
MDL Number: MFCD00047955
Molecular Formula: C5H13NO
Molecular Weight: 103.17
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 126
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: 2-Aminoethyl Propyl Ether, O-Propylethanolamine

Formula: C₅H₁₃NO
MW: 103.17 g/mol
Boiling Pt: 126 °C
Density: 0.85 (20 °C)
MDL Number: MFCD00047955
CAS Number: 42185-03-5
https://us.vwr.com/stibo/web/std.lang.all/92/37/28929237.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR