Phloxin B

Supplier: TCI America

Synonyms: PhloxineEosine Blue2′,4′,5′,7′-Tetrabromo-4,5,6,7-tetrachlorofluorescein

F0142-25G
TCF0142-025GEA 142.1 USD
TCF0142-025G
Phloxin B
Phloxin B

CAS Number: 18472-87-2
MDL Number: MFCD00070627
Molecular Formula: C20H4Br4Cl4O5
Molecular Weight: 829.63
Form: Powder
Color: Red

Formula: C₂₀H₂Br₄Cl₄Na₂O₅
MW: 829.6344 g/mol
MDL Number: MFCD00070627
CAS Number: 18472-87-2
https://us.vwr.com/stibo/web/std.lang.all/87/58/28968758.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR