Furoin ≥96.0%
Supplier: TCI America
Synonyms:
2,2-Furoin
F0082-5G
F0082-25G
TCF0082-025GEA
350
USD
TCF0082-025G
TCF0082-5G
Furoin ≥96.0%
Furoin
CAS Number: 552-86-3
MDL Number: MFCD00003246
Molecular Formula: C10H8O4
Molecular Weight: 192.17
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 139
Formula:
C₁₀H₈O₄ MW: 192.17 g/mol Melting Pt: 139 °C |
MDL Number:
MFCD00003246 CAS Number: 552-86-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...