Flavianic acid dihydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2,4-Dinitro-1-naphthol-7-sulphonic acid
, 8-Hydroxy-5,7-dinitro-2-naphthalenesulphonic acid
, 8-Hydroxy-5,7-dinitronaphthalene-2-sulphonic acid
, 2,4-Dinitro-1-naphthol-7-sulfonic acid
, 8-Hydroxy-5,7-dinitronaphthalene-2-sulfonic acid
, 8-Hydroxy-5,7-dinitro-2-naphthalenesulfonic acid
CAS Number: 483-84-1
MDL Number: MFCD00046407
Molecular Formula: C10H6N2O8S
Molecular Weight: 314.22
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 149
Other Product Names / Synonyms: 2,4-Dinitro-1-naphthol-7-sulfonic Acid, 8-Hydroxy-5,7-dinitro-2-naphthalenesulfonic Acid
Formula:
C₁₀H₆N₂O₈S MW: 314.22 g/mol Melting Pt: 149 °C |
MDL Number:
MFCD00046407 CAS Number: 483-84-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...