TCIAF0003

F0003-25ML
TCF0003-025MLEA 11.13 USD
TCF0003-025ML
TCIAF0003
CAS Number: 8006-84-6
MDL Number: MFCD00146918
Purity/Analysis Method: 80.0% (GC)
Form: Clear Liquid
Flash Point (°C): 31
Freezing point (°C): 15
Specific Gravity (20/20): 0.98
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR