1,2-Epoxypentan ≥98.0%

Supplier: TCI America

Synonyms: 1,2-EpoxypentanePropyloxirane1,2-Pentylene Oxide

E0822-5ML E0822-25ML
TCE0822-25MLEA 409 USD
TCE0822-25ML TCE0822-5ML
1,2-Epoxypentan ≥98.0%
1,2-Epoxypentan

CAS Number: 1003-14-1
MDL Number: MFCD00192124
Molecular Formula: C5H10O
Molecular Weight: 86.13
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): -12
Specific Gravity (20/20): 0.83


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2-Pentylene Oxide, Propyloxirane

https://us.vwr.com/stibo/web/std.lang.all/82/17/28968217.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR