2-Ethoxy-5-(1-propenyl)phenol ≥98.0%

Supplier: TCI America

Synonyms: 1-Ethoxy-2-hydroxy-4-(1-propenyl)benzenePropenylguaethol

E0804-100G E0804-25G
TCE0804-25GEA 68 USD
TCE0804-25G TCE0804-100G
2-Ethoxy-5-(1-propenyl)phenol ≥98.0%
2-Ethoxy-5-(1-propenyl)phenol

CAS Number: 94-86-0
MDL Number: MFCD00009283
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 116
Melting point (°C): 86
Flash Point (°C): 100


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: Propenylguaethol, 1-Ethoxy-2-hydroxy-4-(1-propenyl)benzene

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 116 °C
Melting Pt: 86 °C
Flash Pt: 100 °C
MDL Number: MFCD00009283
CAS Number: 94-86-0
https://us.vwr.com/stibo/web/std.lang.all/82/04/28968204.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR