DL-Alanylglycylglycine dihydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: H-DL-Ala-Gly-Gly-OH

A0184-100MG
TCA0184-100MGEA 88 USD
TCA0184-100MG
DL-Alanylglycylglycine dihydrate ≥98.0% (by titrimetric analysis)
DL-Alanylglycylglycine dihydrate

CAS Number: 927-21-9
MDL Number: MFCD00270471
Molecular Formula: C7H13N3O4
Molecular Weight: 203.20
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 198
Storage Temperature: 0-10°C


Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-DL-Ala-Gly-Gly-OH

Formula: C₇H₁₃N₃O₄
MW: 203.20 g/mol
Melting Pt: 198 °C
MDL Number: MFCD00270471
CAS Number: 927-21-9
https://us.vwr.com/stibo/web/std.lang.all/76/33/28927633.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR