Ethyl-2,4,6-trimethylbenzoate ≥95.0%
Supplier: TCI America
Synonyms:
Ethyl 2,4,6-trimethylbenzoate
, Ethyl Mesitylenecarboxylate
, Mesitylenecarboxylic Acid Ethyl Ester
, 2,4,6-Trimethylbenzoic Acid Ethyl Ester
E0736-25G
TCE0736-25GEA
393
USD
TCE0736-25G
Ethyl-2,4,6-trimethylbenzoate ≥95.0%
Ethyl-2,4,6-trimethylbenzoate
CAS Number: 1754-55-8
MDL Number: MFCD00015439
Molecular Formula: C12H16O2
Molecular Weight: 192.26
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 121
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: 2,4,6-Trimethylbenzoic Acid Ethyl Ester, Ethyl Mesitylenecarboxylate, Mesitylenecarboxylic Acid Ethyl Ester
Formula:
C₁₂H₁₆O₂ MW: 192.26 g/mol Boiling Pt: 121 °C Density: 1.00 (20 °C) |
MDL Number:
MFCD00015439 CAS Number: 1754-55-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...