3-Ethoxythiophenol ≥98.0%
Supplier: TCI America
E0701-5G
E0701-1G
TCE0701-1GEA
113.63
USD
TCE0701-1G
TCE0701-5G
3-Ethoxythiophenol ≥98.0%
3-Ethoxythiophenol
CAS Number: 86704-82-7
MDL Number: MFCD03094660
Molecular Formula: C8H10OS
Molecular Weight: 154.23
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 239
Flash Point (°C): 110
Specific Gravity (20/20): 1.10
Other Product Names / Synonyms: 3-Ethoxythiophenol
Formula:
C₈H₁₀OS Boiling Pt: 239 °C Density: 1.10 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD03094660 CAS Number: 86704-82-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...