Ethionamide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Ethylpyridine-4-carbothioamide2-Ethylthioisonicotinamide

E0695-5G E0695-25G
TCE0695-5GEA 36.94 USD
TCE0695-5G TCE0695-25G
Ethionamide ≥98.0% (by titrimetric analysis)
Ethionamide

CAS Number: 536-33-4
MDL Number: MFCD00057361
Molecular Formula: C8H10N2S
Molecular Weight: 166.24
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Boiling point (°C): 167
Melting point (°C): 162


Product Categories: Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: 2-Ethylpyridine-4-carbothioamide, 2-Ethylthioisonicotinamide

Formula: C₈H₁₀N₂S
MW: 166.24 g/mol
Boiling Pt: 167 °C
Melting Pt: 162 °C
MDL Number: MFCD00057361
CAS Number: 536-33-4
https://us.vwr.com/stibo/web/std.lang.all/81/15/28968115.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR