Ethyl-6-methyl-2-oxo-3-cyclohexene-1-carboxylate (mixture of isomers) ≥95.0%

Supplier: TCI America

Synonyms: Ethyl 3-Methyl-5-cyclohexen-1-one-2-carboxylate6-Methyl-2-oxo-3-cyclohexene-1-carboxylic Acid Ethyl Ester

E0646-5G
TCE0646-5GEA 291.24 USD
TCE0646-5G
Ethyl-6-methyl-2-oxo-3-cyclohexene-1-carboxylate (mixture of isomers) ≥95.0%
Ethyl-6-methyl-2-oxo-3-cyclohexene-1-carboxylate (mixture of isomers)

CAS Number: 3419-32-7
Molecular Formula: C10H14O3
Molecular Weight: 182.22
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: Ethyl 3-Methyl-5-cyclohexen-1-one-2-carboxylate, 6-Methyl-2-oxo-3-cyclohexene-1-carboxylic Acid Ethyl Ester

Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
Boiling Pt: 110 °C
Density: 1.07 (20 °C)
CAS Number: 3419-32-7
https://us.vwr.com/stibo/web/std.lang.all/80/84/28968084.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR