9-Ethylcarbazole-3-carboxaldehyde-N-ethyl-N-phenylhydrazone ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 9-Ethyl-3-(N-ethyl-N-phenylhydrazonomethyl)carbazole

E0576-1G
TCE0576-1GEA 389 USD
TCE0576-1G
9-Ethylcarbazole-3-carboxaldehyde-N-ethyl-N-phenylhydrazone ≥98.0% (by HPLC)
9-Ethylcarbazole-3-carboxaldehyde-N-ethyl-N-phenylhydrazone

CAS Number: 84678-52-4
MDL Number: MFCD03208617
Molecular Formula: C23H23N3
Molecular Weight: 341.46
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 9-Ethyl-3-(N-ethyl-N-phenylhydrazonomethyl)carbazole

Formula: C₂₃H₂₃N₃
MW: 341.46 g/mol
MDL Number: MFCD03208617
CAS Number: 84678-52-4
https://us.vwr.com/stibo/web/std.lang.all/80/69/28968069.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR