9-Ethylcarbazole-3-carboxaldehyde-N-butyl-N-phenylhydrazone ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 9-Ethyl-3-(N-butyl-N-phenylhydrazonomethyl)carbazole

E0575-1G
TCE0575-1GEA 272 USD
TCE0575-1G
9-Ethylcarbazole-3-carboxaldehyde-N-butyl-N-phenylhydrazone ≥98.0% (by HPLC)
9-Ethylcarbazole-3-carboxaldehyde-N-butyl-N-phenylhydrazone

CAS Number: 88107-84-0
MDL Number: MFCD06797101
Molecular Formula: C25H27N3
Molecular Weight: 369.51
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 9-Ethyl-3-(N-butyl-N-phenylhydrazonomethyl)carbazole

Formula: C₂₅H₂₇N₃
MW: 369.51 g/mol
MDL Number: MFCD06797101
CAS Number: 88107-84-0
https://us.vwr.com/stibo/web/std.lang.all/80/67/28968067.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR