9-Ethylcarbazole-3-carboxaldehyde diphenylhydrazone ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 3-(Diphenylhydrazonomethyl)-9-ethylcarbazole

E0571-1G
TCE0571-1GEA 201.89 USD
TCE0571-1G
9-Ethylcarbazole-3-carboxaldehyde diphenylhydrazone ≥98.0% (by HPLC)
9-Ethylcarbazole-3-carboxaldehyde diphenylhydrazone

CAS Number: 73276-70-7
MDL Number: MFCD00144879
Molecular Formula: C27H23N3
Molecular Weight: 389.50
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 159


Other Product Names / Synonyms: 3-(Diphenylhydrazonomethyl)-9-ethylcarbazole

Formula: C₂₇H₂₃N₃
MW: 389.50 g/mol
Melting Pt: 159 °C
MDL Number: MFCD00144879
CAS Number: 73276-70-7
https://us.vwr.com/stibo/web/std.lang.all/80/64/28968064.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR