TCIAE0529

E0529-25ML
TCE0529-025MLEA 112.42 USD
TCE0529-025ML
TCIAE0529
CAS Number: 103-75-3
MDL Number: MFCD00006561
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 24
Specific Gravity (20/20): 0.97
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR