1,2-Epoxycyclooctane ≥97.0%
Supplier: TCI America
E0516-25G
E0516-5G
TCE0516-005GEA
79.04
USD
TCE0516-005G
TCE0516-025G
1,2-Epoxycyclooctane ≥97.0%
1,2-Epoxycyclooctane
CAS Number: 286-62-4
MDL Number: MFCD00005164
Molecular Formula: C8H14O
Molecular Weight: 126.20
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 75
Melting point (°C): 55
Flash Point (°C): 56
Other Product Names / Synonyms: Cyclooctene Oxide, 9-Oxabicyclo[6.1.0]nonane
Formula:
C₈H₁₄O Boiling Pt: 75 °C Melting Pt: 55 °C Flash Pt: 56 °C |
MDL Number:
MFCD00005164 CAS Number: 286-62-4 UN: 1325 ADR: 4.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...