Ethylene glycol mono(4-bromophenyl)ether ≥98.0%

Supplier: TCI America

Synonyms: 2-(4-Bromophenoxy)ethanol

E0504-25G
TCE0504-025GEA 323.79 USD
TCE0504-025G
Ethylene glycol mono(4-bromophenyl)ether ≥98.0%
Ethylene glycol mono(4-bromophenyl)ether

CAS Number: 34743-88-9
MDL Number: MFCD00002860
Molecular Formula: C8H9BrO2
Molecular Weight: 217.06
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 184
Melting point (°C): 55
Flash Point (°C): 110


Other Product Names / Synonyms: 2-(4-Bromophenoxy)ethanol

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Boiling Pt: 184 °C
Melting Pt: 55 °C
Flash Pt: 110 °C
MDL Number: MFCD00002860
CAS Number: 34743-88-9
https://us.vwr.com/stibo/web/std.lang.all/80/15/28968015.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR