TCIAA1294

A1294-25G
TCA1294-025GEA 201.93 USD
TCA1294-025G
TCIAA1294
CAS Number: 16407-55-9
MDL Number: MFCD00191299
Molecular Formula: C10H10BrNS
Molecular Weight: 256.16
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 150
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR