Ethylenedioxybis(trimethylsilane) ≥97.0%

Supplier: TCI America

Synonyms: 1,2-Bis(trimethylsilyloxy)ethaneEthylene Glycol Bis(trimethylsilyl Ether)

E0478-25G
TCE0478-025GEA 61.92 USD
TCE0478-025G
Ethylenedioxybis(trimethylsilane) ≥97.0%
Ethylenedioxybis(trimethylsilane)

[Protecting Reagent for Aldehydes and Ketones]
CAS Number: 7381-30-8
MDL Number: MFCD00009640
Molecular Formula: C8H22O2Si2
Molecular Weight: 206.43
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 166
Flash Point (°C): 46
Specific Gravity (20/20): 0.84


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: 1,2-Bis(trimethylsilyloxy)ethane, Ethylene Glycol Bis(trimethylsilyl Ether)

Formula: C₈H₂₂O₂Si₂
MW: 206.43 g/mol
Boiling Pt: 166 °C
Density: 0.84 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00009640
CAS Number: 7381-30-8
https://us.vwr.com/stibo/web/std.lang.all/79/93/28967993.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR