DL-Alanylglycine

Supplier: TCI
A0183-1G
TCA0183-001GEA 143 USD
TCA0183-001G
DL-Alanylglycine
Proteins and Peptides
  • Melting point: 225 °C
  • Solubility in water: Almost transparency

Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-DL-Ala-Gly-OH
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR