Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4'-Ethylpropiophenone

Supplier: TCI America


E0461-25ML E0461-500ML
TCE0461-025MLEA 70 USD InStock
TCE0461-025ML TCE0461-500ML
4'-Ethylpropiophenone
4'-Ethylpropiophenone
CAS Number: 27465-51-6
MDL Number: MFCD00191646
Molecular Formula: C11H14O
Molecular Weight: 162.23
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 246
Specific Gravity (20/20): 0.98

Other Product Names / Synonyms: Ethyl 4-Ethylphenyl Ketone


Formula: C₁₁H₁₄O
Boiling Pt: 246 °C
Density: 0,98 (20 °C)
MDL Number: MFCD00191646
CAS Number: 27465-51-6
EINECS: 000-000-0
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty