D-Erythronolactone ≥98.0%
Supplier: TCI America
Synonyms:
(3R,4R)-Dihydroxydihydro-2(3H)-furanone
CAS Number: 15667-21-7
MDL Number: MFCD00077763
Molecular Formula: C4H6O4
Molecular Weight: 118.09
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 103
Specific rotation [a]20/D: -71 deg (C=1, H2O)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Sugar Acids, Erythroses
Other Product Names / Synonyms: (3R,4R)-Dihydroxydihydro-2(3H)-furanone, D-Erythronic gamma-Lactone
Formula:
C₄H₆O₄ MW: 118.09 g/mol Melting Pt: 103 °C |
MDL Number:
MFCD00077763 CAS Number: 15667-21-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...