(3S)-(-)-3-(Ethylamino)pyrrolidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0434-1G
TCE0434-001GEA 467 USD
TCE0434-001G
(3S)-(-)-3-(Ethylamino)pyrrolidine ≥98.0% (by GC, titration analysis)
(3S)-(-)-3-(Ethylamino)pyrrolidine

CAS Number: 381670-31-1
MDL Number: MFCD00191359
Molecular Formula: C6H14N2
Molecular Weight: 114.19
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 176
Specific Gravity (20/20): 0.92
Specific rotation [a]20/D: -15 deg (C=10, EtOH)

Formula: C₆H₁₄N₂
Boiling Pt: 176 °C
Density: 0.92 (20 °C)
MDL Number: MFCD00191359
CAS Number: 381670-31-1
https://us.vwr.com/stibo/web/std.lang.all/79/51/28967951.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR