Elaidic anhydride

Synonyms: trans-9-Octadecenoic Anhydride

E0418-5G E0418-1G
TCE0418-001GEA 120.36 USD
TCE0418-001G TCE0418-005G
Elaidic anhydride
Elaidic anhydride

CAS Number: 55726-25-5
MDL Number: MFCD00070026
Molecular Formula: C36H66O3
Molecular Weight: 546.92
Form: Crystal
Color: White
Melting point (°C): 46


Other Product Names / Synonyms: trans-9-Octadecenoic Anhydride

Formula: C₃₆H₆₆O₃
MW: 546.92 g/mol
Melting Pt: 46 °C
MDL Number: MFCD00070026
CAS Number: 55726-25-5
https://us.vwr.com/stibo/web/std.lang.all/79/36/28967936.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR