Ethyleneglycolbis[4-(ethoxycarbonyl)phenyl]ether ≥98.0%

Supplier: TCI America

Synonyms: 1,2-Bis(4-carbethoxyphenoxy)ethane1,2-Bis(4-ethoxycarbonylphenoxy)ethaneEthylene Glycol Bis(4-carbethoxyphenyl) Ether

E0414-5G
TCE0414-005GEA 99.88 USD
TCE0414-005G
Ethyleneglycolbis[4-(ethoxycarbonyl)phenyl]ether ≥98.0%
Ethyleneglycolbis[4-(ethoxycarbonyl)phenyl]ether

CAS Number: 25909-66-4
MDL Number: MFCD00144288
Molecular Formula: C20H22O6
Molecular Weight: 358.39
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 108


Other Product Names / Synonyms: 1,2-Bis(4-carbethoxyphenoxy)ethane, 1,2-Bis(4-ethoxycarbonylphenoxy)ethane, Ethylene Glycol Bis(4-carbethoxyphenyl) Ether

Formula: C₂₀H₂₂O₆
MW: 358.39 g/mol
Melting Pt: 108 °C
MDL Number: MFCD00144288
CAS Number: 25909-66-4
https://us.vwr.com/stibo/web/std.lang.all/79/32/28967932.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR