1-(2-Ethoxyphenyl)piperazine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

E0408-25G
TCE0408-025GEA 157 USD
TCE0408-025G
1-(2-Ethoxyphenyl)piperazine hydrochloride ≥98.0% (by HPLC, titration analysis)
1-(2-Ethoxyphenyl)piperazine hydrochloride

CAS Number: 83081-75-8
MDL Number: MFCD00012763
Molecular Formula: C12H18N2O
Molecular Weight: 242.75
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 207

Formula: C₁₂H₁₈N₂O
Melting Pt: 207 °C
MDL Number: MFCD00012763
CAS Number: 83081-75-8
https://us.vwr.com/stibo/web/std.lang.all/79/27/28967927.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR