Allylsuccinic anhydride ≥97.0%

Supplier: TCI America

Synonyms: 4-Pentene-1,2-dicarboxylic Anhydride(2-Propenyl)butanedioic Anhydride

A1283-25G A1283-5G
TCA1283-025GEA 198.94 USD
TCA1283-025G TCA1283-005G
Allylsuccinic anhydride ≥97.0%
Allylsuccinic anhydride

CAS Number: 7539-12-0
MDL Number: MFCD00085227
Molecular Formula: C7H8O3
Molecular Weight: 140.14
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 143
Specific Gravity (20/20): 1.17


Product Categories: Materials Chemistry, Monomers, Monomers
Other Product Names / Synonyms: 4-Pentene-1,2-dicarboxylic Anhydride, (2-Propenyl)butanedioic Anhydride

Formula: C₇H₈O₃
MW: 140.14 g/mol
Boiling Pt: 143 °C
Density: 1.17 (20 °C)
MDL Number: MFCD00085227
CAS Number: 7539-12-0
https://us.vwr.com/stibo/web/std.lang.all/91/73/28929173.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR