Ethyl-2-ethylbutyrate ≥99.0%
Supplier: TCI America
Synonyms:
Ethyl 2-ethylbutanoate
E0345-500ML
E0345-25ML
TCE0345-025MLEA
91.31
USD
TCE0345-025ML
TCE0345-500ML
Ethyl-2-ethylbutyrate ≥99.0%
Ethyl 2-ethylbutyrate
CAS Number: 2983-38-2
MDL Number: MFCD00059396
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 152
Flash Point (°C): 39
Specific Gravity (20/20): 0.87
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: 2-Ethylbutyric Acid Ethyl Ester
Formula:
C₈H₁₆O₂ MW: 144.21 g/mol Boiling Pt: 152 °C Density: 0.87 (20 °C) Flash Pt: 39 °C |
MDL Number:
MFCD00059396 CAS Number: 2983-38-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...