1,2-Epoxycyclododecane (mixture of cis and trans isomers) ≥95.0%

Supplier: TCI America

Synonyms: 1,2-Epoxycyclododecane (mixture of isomers)13-Oxabicyclo[10.1.0]tridecaneCyclododecene OxideEpoxycyclododecane

E0322-25ML E0322-5ML
TCE0322-025MLEA 111.18 USD
TCE0322-025ML TCE0322-005ML
1,2-Epoxycyclododecane (mixture of cis and trans isomers) ≥95.0%
1,2-Epoxycyclododecane (mixture of cis and trans isomers)

CAS Number: 286-99-7
MDL Number: MFCD00005116
Molecular Formula: C12H22O
Molecular Weight: 182.31
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 113
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Cyclododecene Oxide, 13-Oxabicyclo[10.1.0]tridecane

Formula: C₁₂H₂₂O
Boiling Pt: 130 °C
Density: 0.95 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00005116
CAS Number: 286-99-7
https://us.vwr.com/stibo/web/std.lang.all/78/54/28967854.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR