1,2-Epoxyheptane ≥96.0%

Supplier: TCI America

E0312-5ML
TCE0312-005MLEA 320 USD
TCE0312-005ML
1,2-Epoxyheptane ≥96.0%
1,2-Epoxyheptane

CAS Number: 5063-65-0
MDL Number: MFCD00037521
Molecular Formula: C7H14O
Molecular Weight: 114.19
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 144
Specific Gravity (20/20): 0.84


Product Categories: Materials Chemistry, Monomers, Analytical Science, Odor Deterioration Factors of Cosmetics
Other Product Names / Synonyms: 1,2-Heptylene Oxide

Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 144 °C
Density: 0.84 (20 °C)
CAS Number: 5063-65-0

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR