N-(3-Aminopropyl)diethanolamine ≥90.0%

Supplier: TCI America

Synonyms: N,N-Bis(2-hydroxyethyl)-1,3-diaminopropane

A1280-500G A1280-25G
TCA1280-500GEA 910.18 USD
TCA1280-500G TCA1280-025G
N-(3-Aminopropyl)diethanolamine ≥90.0%
N-(3-Aminopropyl)diethanolamine

CAS Number: 4985-85-7
MDL Number: MFCD00047971
Molecular Formula: C7H18N2O2
Molecular Weight: 162.23
Purity/Analysis Method: 90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 207
Flash Point (°C): 138
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: N,N-Bis(2-hydroxyethyl)-1,3-diaminopropane

Formula: C₇H₁₈N₂O₂
MW: 162.23 g/mol
Boiling Pt: 207 °C
Density: 1.07 (20 °C)
Flash Pt: 138 °C
MDL Number: MFCD00047971
CAS Number: 4985-85-7
https://us.vwr.com/stibo/web/std.lang.all/91/69/28929169.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR