1,2-Epoxyeicosane ≥80.0%
Supplier: TCI America
Synonyms:
1,2-Eicosylene Oxide
E0311-5G
TCE0311-005GEA
279.05
USD
TCE0311-005G
1,2-Epoxyeicosane ≥80.0%
1,2-Epoxyeicosane
CAS Number: 19780-16-6
MDL Number: MFCD00059157
Molecular Formula: C20H40O
Molecular Weight: 296.54
Purity/Analysis Method:
Form: Crystal
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2-Eicosylene Oxide
Formula:
C₂₀H₄₀O MW: 296.54 g/mol |
MDL Number:
MFCD00059157 CAS Number: 19780-16-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...