Ethylenediamine-N,N'-diacetic-N,N'-dipropionic acid hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: EDDADP

E0288-1G
TCE0288-001GEA 161 USD
TCE0288-001G
Ethylenediamine-N,N'-diacetic-N,N'-dipropionic acid hydrate ≥98.0% (by titrimetric analysis)
Ethylenediamine-N,N'-diacetic-N,N'-dipropionic acid hydrate

CAS Number: 32701-19-2
MDL Number: MFCD00002768
Molecular Formula: C12H20N2O8
Molecular Weight: 320.30
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Product Categories: Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: EDDADP

Formula: C₁₂H₂₀N₂O₈
MW: 320.30 g/mol
MDL Number: MFCD00002768
CAS Number: 32701-19-2
https://us.vwr.com/stibo/web/std.lang.all/78/30/28967830.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR