6-Ethoxy-2-methylquinoline ≥98.0% (by GC, titration analysis)
Synonyms:
2-Methyl-6-ethoxyquinoline
E0251-5G
TCE0251-005GEA
286.38
USD
TCE0251-005G
6-Ethoxy-2-methylquinoline ≥98.0% (by GC, titration analysis)
6-Ethoxy-2-methylquinoline
CAS Number: 6628-28-0
MDL Number: MFCD00023987
Molecular Formula: C12H13NO
Molecular Weight: 187.24
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 70
Other Product Names / Synonyms: 6-Ethoxy-2-methylquinoline, 6-Ethoxyquinaldine
Formula:
C₁₂H₁₃NO MW: 187.24 g/mol Melting Pt: 70 °C |
MDL Number:
MFCD00023987 CAS Number: 6628-28-0 EINECS: 229-609-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...