5-Methyl-3-hexanone ≥98.0%
Supplier: TCI America
Synonyms:
Ethyl Isobutyl ketone
, 5-Methyl-3-hexanone
E0249-10ML
TCE0249-010MLEA
270.99
USD
TCE0249-010ML
5-Methyl-3-hexanone ≥98.0%
5-Methyl-3-hexanone
CAS Number: 623-56-3
MDL Number: MFCD00009319
Molecular Formula: C7H14O
Molecular Weight: 114.19
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 39
Specific Gravity (20/20): 0.81
Other Product Names / Synonyms: 5-Methyl-3-hexanone
Formula:
C₇H₁₄O MW: 114.19 g/mol Boiling Pt: 136 °C Density: 0.81 (20 °C) Flash Pt: 39 °C |
MDL Number:
MFCD00009319 CAS Number: 623-56-3 EINECS: 210-801-9 UN: 1224 ADR: 3,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...