TCIAE0243

E0243-500G E0243-25G
TCE0243-500GEA 45.71 USD
TCE0243-500G TCE0243-025G
TCIAE0243
CAS Number: 110-30-5
MDL Number: MFCD00059224
Molecular Formula: C38H76N2O2
Molecular Weight: 593.04
Purity/Analysis Method: 85.0% (GC)
Form: Crystal
Melting point (°C): 144
Flash Point (°C): 287

Other Product Names / Synonyms: N,N'-Ethylenebisstearamide
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR